missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Aspartic acid 4-tert-butyl ester, 98%
CAS: 3057-74-7 | C8H15NO4 | 189.211 g/mol
Supplier: Thermo Scientific Chemicals H5981006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| L-Aspartic acid 4-tert-butyl ester | |
| C8H15NO4 | |
| 4671089 | |
| MXWMFBYWXMXRPD-YFKPBYRVSA-N | |
| (2S)-2-amino-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid | |
| 189.211 | |
| −9° (c=2 in Methanol: Water (4:1)) | |
| 98% |
| 3057-74-7 | |
| MFCD00038577 | |
| h-asp otbu-oh, l-aspartic acid 4-tert-butyl ester, h-asp obut-oh, 4-tert-butyl hydrogen l-aspartate, 2s-2-amino-4-tert-butoxy-4-oxobutanoic acid, s-2-amino-4-tert-butoxy-4-oxobutanoic acid, aspartic acid, 4-tert-butyl ester, asp but, asp obut, asp otbu oh | |
| CC(C)(C)OC(=O)CC(C(=O)O)N | |
| 5 g | |
| 7010570 | |
| 189.21 |
Chemical Identifiers
| 3057-74-7 | |
| 189.211 | |
| MXWMFBYWXMXRPD-YFKPBYRVSA-N | |
| 7010570 | |
| CC(C)(C)OC(=O)CC(C(=O)O)N |
| C8H15NO4 | |
| MFCD00038577 | |
| h-asp otbu-oh, l-aspartic acid 4-tert-butyl ester, h-asp obut-oh, 4-tert-butyl hydrogen l-aspartate, 2s-2-amino-4-tert-butoxy-4-oxobutanoic acid, s-2-amino-4-tert-butoxy-4-oxobutanoic acid, aspartic acid, 4-tert-butyl ester, asp but, asp obut, asp otbu oh | |
| (2S)-2-amino-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid |
Safety and Handling
EINECSNumber : 221-292-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only