missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid 4-methyl ester hydrochloride, 97%

CAS: 16856-13-6 | C5H10ClNO4 | 183.588 g/mol

$70.08 - $233.01

Chemical Identifiers

CAS 16856-13-6
Molecular Formula C5H10ClNO4
Molecular Weight (g/mol) 183.588
MDL Number MFCD00038972
InChI Key QRBMPUYOGOCYDJ-DFWYDOINSA-N
Synonym h-asp ome-oh.hcl, l-aspartic acid beta-methyl ester hydrochloride, s-2-amino-4-methoxy-4-oxobutanoic acid hydrochloride, h-asp ome-oh hcl, 4-methyl l-aspartate hydrochloride, s-2-amino-succinic acid 4-methyl ester hydrochloride, 2s-2-amino-4-methoxy-4-oxobutanoic acid hydrochloride, l-aspartic acid 4-methyl ester hcl, aspartic acid, 4-methylester, hydrochloride, l-aspartic acid, 4-methyl ester, hydrochloride
PubChem CID 12832742
IUPAC Name (2S)-2-amino-4-methoxy-4-oxobutanoic acid;hydrochloride
SMILES COC(=O)CC(C(=O)O)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6217203
View Documents
Thermo Scientific Chemicals
H6217203
1 g
Each for $70.08
Only null left
 
AAH6217206
View Documents
Thermo Scientific Chemicals
H6217206
5 g
Each for $233.01
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16856-13-6
183.588
QRBMPUYOGOCYDJ-DFWYDOINSA-N
12832742
COC(=O)CC(C(=O)O)N.Cl
C5H10ClNO4
MFCD00038972
h-asp ome-oh.hcl, l-aspartic acid beta-methyl ester hydrochloride, s-2-amino-4-methoxy-4-oxobutanoic acid hydrochloride, h-asp ome-oh hcl, 4-methyl l-aspartate hydrochloride, s-2-amino-succinic acid 4-methyl ester hydrochloride, 2s-2-amino-4-methoxy-4-oxobutanoic acid hydrochloride, l-aspartic acid 4-methyl ester hcl, aspartic acid, 4-methylester, hydrochloride, l-aspartic acid, 4-methyl ester, hydrochloride
(2S)-2-amino-4-methoxy-4-oxobutanoic acid;hydrochloride

Specifications

16856-13-6
MFCD00038972
h-asp ome-oh.hcl, l-aspartic acid beta-methyl ester hydrochloride, s-2-amino-4-methoxy-4-oxobutanoic acid hydrochloride, h-asp ome-oh hcl, 4-methyl l-aspartate hydrochloride, s-2-amino-succinic acid 4-methyl ester hydrochloride, 2s-2-amino-4-methoxy-4-oxobutanoic acid hydrochloride, l-aspartic acid 4-methyl ester hcl, aspartic acid, 4-methylester, hydrochloride, l-aspartic acid, 4-methyl ester, hydrochloride
COC(=O)CC(C(=O)O)N.Cl
1 g
12832742
97%
L-Aspartic acid 4-methyl ester hydrochloride
C5H10ClNO4
3628668
QRBMPUYOGOCYDJ-DFWYDOINSA-N
(2S)-2-amino-4-methoxy-4-oxobutanoic acid;hydrochloride
183.588
183.59
Solid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 240-880-5

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only