accessibility menu, dialog, popup

UserName

HClH3COONH2OHO

CAS RN 16856-13-6

IUPAC Name: hydrogen 2-amino-4-methoxy-4-oxobutanoic acid chloride
Synonyms: hydrogen 2-amino-4-methoxy-4-oxobutanoic acid chloride
Molecular Weight (g/mol): 183.59
Molecular Formula: C5H10ClNO4
InChi Key: QRBMPUYOGOCYDJ-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC(=O)CC(N)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)