missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid 4-methyl ester hydrochloride, 97%

Catalog No. AAH6217203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6217203 1 g
AAH6217206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6217203 Supplier Thermo Scientific Chemicals Supplier No. H6217203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 16856-13-6 | C5H10ClNO4 | 183.588 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16856-13-6
Molecular Formula C5H10ClNO4
Molecular Weight (g/mol) 183.588
MDL Number MFCD00038972
InChI Key QRBMPUYOGOCYDJ-DFWYDOINSA-N
Synonym h-asp ome-oh.hcl, l-aspartic acid beta-methyl ester hydrochloride, s-2-amino-4-methoxy-4-oxobutanoic acid hydrochloride, h-asp ome-oh hcl, 4-methyl l-aspartate hydrochloride, s-2-amino-succinic acid 4-methyl ester hydrochloride, 2s-2-amino-4-methoxy-4-oxobutanoic acid hydrochloride, l-aspartic acid 4-methyl ester hcl, aspartic acid, 4-methylester, hydrochloride, l-aspartic acid, 4-methyl ester, hydrochloride
PubChem CID 12832742
IUPAC Name (2S)-2-amino-4-methoxy-4-oxobutanoic acid;hydrochloride
SMILES COC(=O)CC(C(=O)O)N.Cl

Specifications

Beilstein 3628668
Quantity 1 g
Formula Weight 183.59
Percent Purity 97%
Physical Form Solid
Chemical Name or Material L-Aspartic acid 4-methyl ester hydrochloride
EINECSNumber 240-880-5
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only