missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Asparagine, Monohydrate, FCC, 98-101.5%, Spectrum™ Chemical
A1373, 5794-13-8, C4H8N2O3·H2O
$224.06 - $38378.33
Chemical Identifiers
| CAS | 5794-13-8 |
|---|---|
| Molecular Formula | C4H10N2O4 |
| Molecular Weight (g/mol) | 150.13 |
| InChI Key | RBMGJIZCEWRQES-DKWTVANSSA-N |
| IUPAC Name | (2S)-2-amino-3-carbamoylpropanoic acid hydrate |
| SMILES | O.N[C@@H](CC(N)=O)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18603723
|
Spectrum Chemical Mfg Cor
A137325GM |
25 g | Amber Glass Bottle |
Each for $224.06
|
|
||||
|
18603724
|
Spectrum Chemical Mfg Cor
A1373100GM |
100 g | Amber Glass Bottle |
Each for $416.41
|
|
||||
|
18603725
|
Spectrum Chemical Mfg Cor
A13731KG |
1 kg | Amber Glass Bottle |
Each for $1,853.39
|
|
||||
|
18603726
|
Spectrum Chemical Mfg Cor
A137325KGBL |
25 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical L-Asparagine, Monohydrate, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 5794-13-8 | |
| 150.13 | |
| (2S)-2-amino-3-carbamoylpropanoic acid hydrate |
| C4H10N2O4 | |
| RBMGJIZCEWRQES-DKWTVANSSA-N | |
| O.N[C@@H](CC(N)=O)C(O)=O |
Specifications
| 5794-13-8 | |
| 0.1% | |
| Amber Glass Bottle | |
| 25 g | |
| +33.0° to +36.5° | |
| (2S)-2-amino-3-carbamoylpropanoic acid hydrate | |
| 98 to 101.5% |
| 100% | |
| 11.5-12.5% | |
| C4H10N2O4 | |
| RBMGJIZCEWRQES-DKWTVANSSA-N | |
| O.N[C@@H](CC(N)=O)C(O)=O | |
| 150.13 | |
| FCC |