missing translation for 'onlineSavingsMsg'
Learn More

L-Asparagine, Monohydrate, FCC, 98-101.5%, Spectrum™ Chemical

Catalog No. 18603726
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603726 25 kg Amber Glass Bottle
18603723 25 g Amber Glass Bottle
18603724 100 g Amber Glass Bottle
18603725 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18603726 Supplier Spectrum Chemical Mfg Cor Supplier No. A137325KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A1373, 5794-13-8, C4H8N2O3·H2O

Spectrum™ Chemical L-Asparagine, Monohydrate, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5794-13-8
Molecular Formula C4H10N2O4
Molecular Weight (g/mol) 150.13
InChI Key RBMGJIZCEWRQES-DKWTVANSSA-N
IUPAC Name (2S)-2-amino-3-carbamoylpropanoic acid hydrate
SMILES O.N[C@@H](CC(N)=O)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 11.5-12.5%
Packaging Amber Glass Bottle
Quantity 25 kg
Optical Rotation +33.0° to +36.5°
Percent Purity 98 to 101.5%
Grade FCC
Physical Form Crystals or Flakes
Chemical Name or Material L-Asparagine, Monohydrate