missing translation for 'onlineSavingsMsg'
Learn More

L-Asparagine, 99%

Catalog No. p-3734457
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC371601000 100 g Glass Bottle
AC371605000 500 g Glass Bottle
AC371600025 2.5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC371601000 Supplier Thermo Scientific Chemicals Supplier No. 371601000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 70-47-3 | C4H8N2O3 | 132.119 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 70-47-3
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.119
MDL Number MFCD00064401
InChI Key DCXYFEDJOCDNAF-REOHCLBHSA-N
Synonym l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid
PubChem CID 6267
ChEBI CHEBI:17196
IUPAC Name (2S)-2,4-diamino-4-oxobutanoic acid
SMILES C(C(C(=O)O)N)C(=O)N

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula H2NCOCH2CH(NH2)CO2H
Specific Rotation + 34.50
Solubility Information Solubility in water: 20g/l (20°C). Other solubilities: insoluble in organic solvents
Water 1% max.
Specific Rotation Condition + 34.50 (20.00°C c=10,2N HCl)
Quantity 100 g
Formula Weight 132.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material L-Asparagine
Show More Show Less

RUO – Research Use Only