missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-(+)-Asparagine, 99%

CAS: 70-47-3 | C4H8N2O3 | 132.119 g/mol

$145.64 - $1519.93

Chemical Identifiers

CAS 70-47-3
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.119
MDL Number MFCD00064401
InChI Key DCXYFEDJOCDNAF-REOHCLBHSA-N
Synonym l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid
PubChem CID 6267
ChEBI CHEBI:17196
IUPAC Name (2S)-2,4-diamino-4-oxobutanoic acid
SMILES C(C(C(=O)O)N)C(=O)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2147322
View Documents
Thermo Scientific Chemicals
B2147322
100 g
Each for $145.64
Only null left
 
AAB2147336
View Documents
Thermo Scientific Chemicals
B2147336
500 g
Each for $414.85
Only null left
 
AAB214730E
View Documents
Thermo Scientific Chemicals
B214730E
2500 g
Each for $1,519.93
Only null left
 
Description

Description

L-(+)-Asparagine acts as a precursor to aspartate. It is also used as a diuretic. It is also employed in the biosynthesis of glycoproteins and proteins. It acts as a food additive. It reacts with reducing sugar to yield acrylamide, which finds application in french fries, potato chips, and toasted bread. It plays an important role in the function and development of the brain. Further, it is useful for human body, physically and mentally. In addition to this, it is used in cell culture media and is a component of (2-methoxyethoxy)methyl (MEM) non-essential amino acid solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(+)-Asparagine acts as a precursor to aspartate. It is also used as a diuretic. It is also employed in the biosynthesis of glycoproteins and proteins. It acts as a food additive. It reacts with reducing sugar to yield acrylamide, which finds application in french fries, potato chips, and toasted bread. It plays an important role in the function and development of the brain. Further, it is useful for human body, physically and mentally. In addition to this, it is used in cell culture media and is a component of (2-methoxyethoxy)methyl (MEM) non-essential amino acid solution.

Solubility
Soluble in acids, bases. Insoluble in methanol, ethanol, ether and benzene.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

70-47-3
132.119
DCXYFEDJOCDNAF-REOHCLBHSA-N
6267
(2S)-2,4-diamino-4-oxobutanoic acid
C4H8N2O3
MFCD00064401
l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid
CHEBI:17196
C(C(C(=O)O)N)C(=O)N

Specifications

234°C to 236°C (decomposition)
C4H8N2O3
1723527
l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid
DCXYFEDJOCDNAF-REOHCLBHSA-N
(2S)-2,4-diamino-4-oxobutanoic acid
132.119
CHEBI:17196
Hygroscopic
L-(+)-Asparagine
70-47-3
MFCD00064401
14,837
Soluble in acids,bases. Insoluble in methanol,ethanol,ether and benzene.
C(C(C(=O)O)N)C(=O)N
100 g
6267
132.12
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-735-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only