missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic Acid (White Crystalline Powder), Fisher BioReagents

Catalog No. BP351500
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP351500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP351500 Supplier Fisher BioReagents Supplier No. FLBP351 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ascorbic Acid is suitable for use in tissue culture systems requiring additives.

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

Alpha Vector Ascorbic Acid
Melting Point 190°C
Color Yellow
Density 1.65g/cm³
Ignition Residue 0.1% max.
Assay Percent Range ≥99.0 %
Packaging Glass Bottle
Quantity 500 g
Merck Index 15, 819
Specific Rotation 20.5 to 21.5° (+ or -)
Formula Weight 176.13
Percent Purity ≥99.0%
Physical Form Solid
Chemical Name or Material L-Ascorbic Acid
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT