missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic Acid, TraceSELECT™, ≥99.0% (RT), Honeywell™ Fluka™

Catalog No. 60044575
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60044575 100 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60044575 Supplier Honeywell Chemicals Supplier No. 05878100G
Only null left

TraceSELECT™,≥99.9998% (metals basis)

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

Melting Point 190°C to 194°C
pH 1.0 to 2.0 (176g/L at 25°C)
Packaging Poly bottle
Quantity 100 g
UN Number NONH for all modes of transport
Beilstein 84272
Solubility Information 176g/L at 20°C
Formula Weight 176.12g/mol
Percent Purity ≥99.0% (RT)
Grade TraceSELECT
Chemical Name or Material L-Ascorbic Acid
Show More Show Less
EINECSNumber 200-066-2
RTECSNumber CI7650000