missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Ascorbic Acid 99.0+%, TCI America™
$37.29 - $85.75
Chemical Identifiers
| CAS | 50-81-7 |
|---|---|
| Molecular Formula | C6H8O6 |
| Molecular Weight (g/mol) | 176.12 |
| MDL Number | MFCD00064328 |
| InChI Key | CIWBSHSKHKDKBQ-DOMZIZNONA-N |
| Synonym | l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin |
| PubChem CID | 54670067 |
| ChEBI | CHEBI:29073 |
| IUPAC Name | (5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one |
| SMILES | OC[C@H](O)[C@H]1OC(=O)C(O)=C1O |
Chemical Identifiers
| 50-81-7 | |
| 176.12 | |
| CIWBSHSKHKDKBQ-DOMZIZNONA-N | |
| 54670067 | |
| (5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one |
| C6H8O6 | |
| MFCD00064328 | |
| l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin | |
| CHEBI:29073 | |
| OC[C@H](O)[C@H]1OC(=O)C(O)=C1O |
Specifications
| 50-81-7 | |
| White | |
| MFCD00064328 | |
| l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin | |
| OC[C@H](O)[C@H]1OC(=O)C(O)=C1O | |
| 176.12 | |
| CHEBI:29073 | |
| ≥99.0% (T) | |
| L-Ascorbic Acid |
| 192°C | |
| C6H8O6 | |
| 25 g | |
| CIWBSHSKHKDKBQ-DOMZIZNONA-N | |
| (5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one | |
| 54670067 | |
| 176.12 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-0062
RTECSNumber : CI7650000
TSCA : Yes