missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic Acid 99.0+%, TCI America™

Catalog No. A053725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A053725G 25 g
A0537500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A053725G Supplier TCI America Supplier No. A053725G
Only null left

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
IUPAC Name (5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

Melting Point 192°C
Color White
Quantity 25 g
Formula Weight 176.12
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Ascorbic Acid
EINECSNumber (5)-0062
RTECSNumber CI7650000
TSCA Yes