missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Ascorbic acid sodium salt, 99%

CAS: 134-03-2 | C6H7NaO6 | 198.11 g/mol

Supplier:  Thermo Scientific Chemicals 352680025

 View more versions of this product

Catalog No. AC352680025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L-Ascorbic acid sodium salt
134-03-2
White to Yellow
98.5 to 101.5% (Iodimetry)
C6H7NaO6
2.5 kg
+104° (25°C c=1,H2O)
sodium ascorbate, l-ascorbic acid sodium salt, sodium l-ascorbate, vitamin c sodium, ascorbicin, sodascorbate, cebitate, aminofenitrooxon, natrii ascorbas, monosodium l-ascorbate
IFVCRSPJFHGFCG-HXPAKLQESA-N
sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
131674100
99%
99%
219.0°C to 221.0°C
Authentic
Plastic bottle
MFCD00082340
15,819
+ 104.00
(5% in water) Clear colorless to light yellow
[Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O
198.11
198.11
Crystalline Powder

Chemical Identifiers

134-03-2
198.11
IFVCRSPJFHGFCG-HXPAKLQESA-N
131674100
[Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O
C6H7NaO6
MFCD00082340
sodium ascorbate, l-ascorbic acid sodium salt, sodium l-ascorbate, vitamin c sodium, ascorbicin, sodascorbate, cebitate, aminofenitrooxon, natrii ascorbas, monosodium l-ascorbate
sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-126-1

RTECSNumber : CI7671000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only