missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Ascorbic Acid, Powder, BAKER ANALYZED™ Biochemical Reagent, J.T. Baker™

Catalog No. p-5391067
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003148 100 g Poly Bottle
02003149 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02003148 Supplier Avantor Supplier No. B58105
Only null left

High quality biochemicals for research use

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

Packaging Poly Bottle
Quantity 100 g
Specific Gravity 1L = 1.65kg
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
Formula Weight 176.12
Grade BAKER ANALYZED™ Biochemical Reagent
Physical Form Powder
Chemical Name or Material L-(+)-Ascorbic Acid