missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic acid, MilliporeSigma™ Supelco™

Catalog No. 111014386
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014386 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014386 Supplier Merck Emd Millipore Supplier No. 47863
Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule) Vitamins, Nutraceuticals and Natural Products

Specifications

Packaging Ampule
CAS 50-81-7
Physical Form Neat
Linear Formula C6H8O6
MDL Number MFCD00064328
pH 1.0 to 2.5 (25°C, 176 g/L aq. soln.)
Melting Point 190°C to 194°C
Synonym L-Threoascorbic acid; Antiscorbutic factor; Vitamin C
Solubility Soluble in water
Type Food Additive Standard
Quantity 1000 mg
Formula Weight 176.12
Grade Analytical Standard
Show More Show Less