missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic Acid (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. p-61088
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A6125 25 g Glass Bottle
A61100 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A6125 Supplier Thermo Fisher Scientific Supplier No. FLA6125
Only null left

Not for human use

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
IUPAC Name (5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

Melting Point 190°C
Color Yellow
Density 1.65g/cm³
Ignition Residue 0.1% max.
Assay Percent Range ≥99.0 %
Packaging Glass Bottle
Linear Formula CH2OHCHOHCHCOH:COHCOO
Quantity 25 g
Specific Rotation +20.5 to +21.5° (+ or -)
Formula Weight 176.13
Percent Purity ≥99.0%
Grade Certified ACS
Physical Form Solid
Chemical Name or Material L-Ascorbic Acid
Show More Show Less