missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L(+)-Ascorbic acid, ACS reagent
CAS: 50-81-7 | C6H8O6 | 176.12 g/mol
Supplier: Thermo Scientific Chemicals 401470025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| L(+)-Ascorbic acid | |
| 50-81-7 | |
| White to Yellow | |
| Authentic | |
| C6H8O6 | |
| 2.5 kg | |
| 15, 819 | |
| + 20.50 | |
| Solubility in water: 333g/L (20°C). Other solubilities: ethanol 20g/L (20°C), soluble methanol and acetone, insoluble in ether, chloroform and benzene | |
| OC[C@H](O)[C@H]1OC(=O)C(O)=C1O | |
| 176.12 | |
| CHEBI:29073 | |
| ≥99.0% | |
| Crystalline Powder |
| 99+% | |
| 190.0°C to 192.0°C | |
| 0.1% max. | |
| Plastic bottle | |
| MFCD00064328 | |
| 18, IV, 3038 | |
| + 20.50 (20.00°C c=10,H2O) | |
| l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin | |
| CIWBSHSKHKDKBQ-DOMZIZNONA-N | |
| (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one | |
| 54670067 | |
| 176.13 | |
| ACS Reagent |
Chemical Identifiers
| 50-81-7 | |
| 176.12 | |
| CIWBSHSKHKDKBQ-DOMZIZNONA-N | |
| 54670067 | |
| (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one |
| C6H8O6 | |
| MFCD00064328 | |
| l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin | |
| CHEBI:29073 | |
| OC[C@H](O)[C@H]1OC(=O)C(O)=C1O |
Safety and Handling
AIR SENSITIVE
LIGHT SENSITIVE
EINECSNumber : 200-066-2
RTECSNumber : CI7650000
TSCA : TSCA
RUO – Research Use Only