missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Ascorbic acid, ACS reagent

Catalog No. AC401471000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC401471000 100 g Plastic bottle
AC401475000 500 g Plastic bottle
AC401470025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC401471000 Supplier Thermo Scientific Chemicals Supplier No. 401471000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 50-81-7 | C6H8O6 | 176.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

Melting Point 190.0°C to 192.0°C
Color White to Yellow
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 100 g
Beilstein 18, IV, 3038
Merck Index 15, 819
Specific Rotation + 20.50
Solubility Information Solubility in water: 333g/L (20°C). Other solubilities: ethanol 20g/L (20°C), soluble methanol and acetone, insoluble in ether, chloroform and benzene
Formula Weight 176.13
Percent Purity ≥99.0%
Grade ACS Reagent
Physical Form Crystalline Powder
Chemical Name or Material L(+)-Ascorbic acid, 99+%
Show More Show Less
Hazard Category LIGHT SENSITIVE
Hazard Statement AIR SENSITIVE
EINECSNumber 200-066-2
RTECSNumber CI7650000
TSCA TSCA

RUO – Research Use Only