missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Ascorbic acid, 99+%

Catalog No. p-7022718
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
2500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1561318 50 g
AAA1561322 100 g
AAA1561336 500 g
AAA156130E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1561318 Supplier Thermo Scientific Chemicals Supplier No. A1561318
Only null left

CAS: 50-81-7 | C6H8O6 | 176.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
L-Ascorbic acid is an all important nutrient and antioxidant. L-ascorbic acid is frequently used as a processing aid or as a preservative in certain nutrients or food components. It is also utilized in the synthetic polymer industry, in photo processing and in metal technology. Miscellaneous uses have been proposed in cosmetics, fibers, preservation of blood, preservation of cut plants, cleaning agents and in assay reagents.

Solubility
Soluble in water. Insoluble in ether, chloroform, benzene, petroleum ether, oils, fats and fat solvents. L-ascorbic acid means that the compound′s stereochemistry is related to that of the levorotatory enantiomer of glyceraldehyde.

Notes
Stable in air when dry.

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

Melting Point 190°C to 193°C (decomposition)
Density 1.65
Quantity 50 g
Beilstein 84272
Merck Index 14,830
Solubility Information Soluble in water. Insoluble in ether,chloroform,benzene,petroleum ether,oils,fats and fat solvents. L-ascorbic acid means that the compound′s stereochemistry is related to that of the levorotatory enantiomer of glyceraldehyde.
Optical Rotation +21° (c=10 in Water)
IUPAC Name (5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
Formula Weight 176.12
Percent Purity ≥99%
Chemical Name or Material L-(+)-Ascorbic acid
Show More Show Less
EINECSNumber 200-066-2
RTECSNumber CI7650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only