missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Ascorbic acid, ≥98%, MP Biomedicals™
Specifications
Specifications
| Color | White to Yellow |
| Density | 1.65g/cm3 |
| Assay Percent Range | ≥98% |
| Quantity | 500 g |
| Formula Weight | 176.1 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | L-Ascorbic acid |
Safety and Handling
| EINECSNumber | 200-066-2 |
| storageNote1 | Protect from light. |
| Recommended Storage | Store at room temperature. |