missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111015427
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015427 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015427 Supplier Merck Emd Millipore Supplier No. PHR11061G
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. L-Arginine is a dibasic, semi-essential amino acid. It acts as a precursor for creatinine and is a natural constituent of most of the dietary proteins. It also plays a key role in many physiological processes such as tissue repair and reproduction.

Specifications

CAS 74-79-3
Physical Form Neat
Linear Formula H2NC(=NH)NH(CH2)3 CH(NH2)CO2H
MDL Number MFCD00002635
Melting Point 222°C (decomposition)
Synonym (S)-2-Amino-5-guanidinopentanoic acid
Quantity 1 g
Formula Weight 174.20
Grade Certified Reference Material