missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine Monohydrochloride, FCC, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18603716
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603716 25 g Poly Bottle
18603717 100 g Poly Bottle
18603718 1 kg Poly Pail
18603719 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18603716 Supplier Spectrum Chemical Mfg Cor Supplier No. A133825GM
Get Up to $500 in Chemicals with Purchase
View all promotions

A1338, 1119-34-2, C6H14N4O2·HCl

Spectrum™ Chemical L-Arginine Monohydrochloride, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1119-34-2
Molecular Formula C6H15ClN4O2
Molecular Weight (g/mol) 210.66
InChI Key KWTQSFXGGICVPE-WCCKRBBISA-N
IUPAC Name hydrogen (2S)-2-amino-5-[(diaminomethylidene)amino]pentanoic acid chloride
SMILES [H+].[Cl-].N[C@@H](CCCN=C(N)N)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Lead (Pb) 5 mg/kg
Loss on Drying 0.3%
Packaging Poly Bottle
Specific Rotation +21.3° to +23.5°
Quantity 25 g
Percent Purity 98.5 to 101.5%
Grade FCC