missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Arginine monohydrochloride, 98+%

Catalog No. AAA1473022
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1473022 100 g
AAA1473036 500 g
AAA147300E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1473022 Supplier Thermo Scientific Chemicals Supplier No. A1473022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

L-Arginine monohydrochloride (L-Arginine hydrochloride or arginine HCl, CAS # 1119-34-2) is the salt of the essential amino acid arginine. | CAS: 1119-34-2 | C6H15ClN4O2 | 210.662 g/mol

L-Arginine monohydrochloride is a precursor of nitric oxide synthesis that induces vasodilation, there by relaxing blood vessels. It is an important intermediate in the urea cycle and involved in the detoxification of nitrogenous wastes. It is commonly used in cell culture media and drug development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • L-Arginine is the left-handed enantiomer of the amino acid arginine, an essential amino acid in juvenile humans
  • Arginine residues are common active site components in enzymes
  • Arginine is involved in nitric acid production, and many biological processes depend on it
  • Arginine plays a role in the immune system and in the detoxification of nitrogenous wastes

Application

  • L-Arginine monohydrochloride is suitable for biochemistry and in vitro research

Chemical Identifiers

CAS 1119-34-2
Molecular Formula C6H15ClN4O2
Molecular Weight (g/mol) 210.662
MDL Number MFCD00064550
InChI Key KWTQSFXGGICVPE-WCCKRBBISA-N
Synonym l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
PubChem CID 66250
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN=C(N)N.Cl

Specifications

Melting Point 216°C (decomposition)
Beilstein 3631658
Merck Index 14,780
Solubility Information Soluble in water. Slightly soluble in hot alcohol.
Density 1.42
Quantity 100 g
Formula Weight 210.67
Optical Rotation +22° (c=5 in 5N HCl)
Percent Purity ≥98%
Sensitivity Hygroscopic
Chemical Name or Material L-Arginine monohydrochloride
Show More Show Less
EINECSNumber 214-275-1
RTECSNumber CF1995500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only