missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Arginine monohydrochloride, 98+%

L-Arginine monohydrochloride (L-Arginine hydrochloride or arginine HCl, CAS # 1119-34-2) is the salt of the essential amino acid arginine. | CAS: 1119-34-2 | C6H15ClN4O2 | 210.662 g/mol

Supplier:  Thermo Scientific Chemicals A147300E

Quantity 2500 g

 View more versions of this product

Catalog No. AAA147300E


Only null left

Description

Description

L-Arginine monohydrochloride is a precursor of nitric oxide synthesis that induces vasodilation, there by relaxing blood vessels. It is an important intermediate in the urea cycle and involved in the detoxification of nitrogenous wastes. It is commonly used in cell culture media and drug development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • L-Arginine is the left-handed enantiomer of the amino acid arginine, an essential amino acid in juvenile humans
  • Arginine residues are common active site components in enzymes
  • Arginine is involved in nitric acid production, and many biological processes depend on it
  • Arginine plays a role in the immune system and in the detoxification of nitrogenous wastes

Application

  • L-Arginine monohydrochloride is suitable for biochemistry and in vitro research
Specifications

Chemical Identifiers

1119-34-2
210.662
KWTQSFXGGICVPE-WCCKRBBISA-N
66250
C(CC(C(=O)O)N)CN=C(N)N.Cl
C6H15ClN4O2
MFCD00064550
l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride

Specifications

216°C (decomposition)
1119-34-2
MFCD00064550
14,780
Soluble in water. Slightly soluble in hot alcohol.
1.42
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
210.662
210.67
≥98%
L-Arginine monohydrochloride
C6H15ClN4O2
3631658
l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
KWTQSFXGGICVPE-WCCKRBBISA-N
C(CC(C(=O)O)N)CN=C(N)N.Cl
2500 g
66250
+22° (c=5 in 5N HCl)
Hygroscopic
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-275-1

RTECSNumber : CF1995500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions