missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Arginine methyl ester dihydrochloride, 98%
CAS: 26340-89-6 | C7H18Cl2N4O2 | 261.147 g/mol
$66.51 - $219.36
Chemical Identifiers
| CAS | 26340-89-6 |
|---|---|
| Molecular Formula | C7H18Cl2N4O2 |
| Molecular Weight (g/mol) | 261.147 |
| MDL Number | MFCD00038948 |
| InChI Key | XQYZOBNLCUAXLF-XRIGFGBMSA-N |
| PubChem CID | 117776 |
| IUPAC Name | methyl (2S)-2-amino-5-(diaminomethylideneamino)pentanoate;dihydrochloride |
| SMILES | COC(=O)C(CCCN=C(N)N)N.Cl.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0595906
|
Thermo Scientific Chemicals
L0595906 |
5 g |
Each for $66.51
|
|
|||||
|
AAL0595914
|
Thermo Scientific Chemicals
L0595914 |
25 g |
Each for $219.36
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 26340-89-6 | |
| 261.147 | |
| XQYZOBNLCUAXLF-XRIGFGBMSA-N | |
| methyl (2S)-2-amino-5-(diaminomethylideneamino)pentanoate;dihydrochloride |
| C7H18Cl2N4O2 | |
| MFCD00038948 | |
| 117776 | |
| COC(=O)C(CCCN=C(N)N)N.Cl.Cl |
Specifications
| ∽190°C (decomposition) | |
| C7H18Cl2N4O2 | |
| 4159929 | |
| XQYZOBNLCUAXLF-XRIGFGBMSA-N | |
| methyl (2S)-2-amino-5-(diaminomethylideneamino)pentanoate;dihydrochloride | |
| 261.147 | |
| +21° (c=2.5 in Methanol) | |
| Hygroscopic | |
| L-Arginine methyl ester dihydrochloride |
| 26340-89-6 | |
| MFCD00038948 | |
| l-arginine methyl ester dihydrochloride, h-arg-ome.2hcl, methyl l-argininate dihydrochloride, h-arg-ome 2hcl, l-arginine, methyl ester, dihydrochloride, s-methyl 2-amino-5-guanidinopentanoate dihydrochloride, methyl 2s-2-amino-5-carbamimidamidopentanoate dihydrochloride, l-arginine, methyl ester, hydrochloride 1:2, h-arg-ome?hcl, h-arg-ome 2 hcl | |
| COC(=O)C(CCCN=C(N)N)N.Cl.Cl | |
| 5 g | |
| 117776 | |
| 261.15 | |
| 98% |
Safety and Handling
EINECSNumber : 247-622-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only