missing translation for 'onlineSavingsMsg'
Learn More

Medisca L - Arginine Hydrochloride USP/EP

Catalog No. 14185670
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185670 25 g
14185636 100 g
14185273 5 kg
14185646 1 kg
14185645 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14185670 Supplier Medisca Supplier No. 050304

L - Arginine Hydrochloride USP/EP is white to almost white and varies between crystals or crystalline powder in texture.

L - Arginine Hydrochloride USP/EP is white to almost white and varies between crystals or crystalline powder in texture. It is also hygroscopic and practically odorless.

Alternative name(s): Arginine hydrochloride

Specifications

Chemical Name or Material L - Arginine Hydrochloride USP/EP
Packaging Jar, 50mL, White, HDPE, Round, 38-400, TE Pano Neck
CAS 1119-34-2
Type Drug Standard
Molecular Weight (g/mol) 210.662
Quantity 25 g
Percent Purity 98.5 to 101.5 %
Grade EP, USP