missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca L - Arginine Hydrochloride USP/EP
Description
L - Arginine Hydrochloride USP/EP is white to almost white and varies between crystals or crystalline powder in texture. It is also hygroscopic and practically odorless.
Alternative name(s): Arginine hydrochloride
Specifications
Specifications
| Chemical Name or Material | L - Arginine Hydrochloride USP/EP |
| Packaging | Jar, 150mL, White, HDPE, Round, 63-400, TE Pano Neck |
| CAS | 1119-34-2 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 210.662 |
| Quantity | 100 g |
| Percent Purity | 98.5 to 101.5 % |
| Grade | EP, USP |