missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine hydrochloride, 98.5 to 101.5%

Catalog No. p-7082813
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ1150036 500 g Plastic Bottle
AAJ11500A1 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ1150036 Supplier Thermo Fisher Scientific Supplier No. J1150036

CAS: 1119-34-2 | C6H15ClN4O2 | 210.662 g/mol

Chemical Identifiers

CAS 1119-34-2
Molecular Formula C6H15ClN4O2
Molecular Weight (g/mol) 210.662
MDL Number MFCD00064550
InChI Key KWTQSFXGGICVPE-WCCKRBBISA-N
Synonym l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
PubChem CID 66250
IUPAC Name (2 S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN=C(N)N.Cl

Specifications

Melting Point 216°C (decomposition)
Packaging Plastic Bottle
Linear Formula NH2C(NH)NH(CH2)3CH(NH2)COOH.HCl
Beilstein 3631658
Density 1.42
Quantity 500 g
Formula Weight 210.66
Percent Purity 98.5 to 101.5%
Specific Gravity 1.42
Physical Form Crystalline Powder
Chemical Name or Material L-Arginine hydrochloride
Show More Show Less
EINECSNumber 214-275-1
RTECSNumber CF1995500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only