missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Arabitol, 99%

Catalog No. AC225861000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC225861000 100 g Glass bottle
AC225860050 5 g Glass bottle
AC225860250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC225861000 Supplier Thermo Scientific Chemicals Supplier No. 225861000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7643-75-6 | C5H12O5 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7643-75-6
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064290
InChI Key HEBKCHPVOIAQTA-UHFFFAOYNA-N
Synonym l-arabitol, l-arabinitol, l-arabinol, l-lyxitol, l---arabitol, arabinitol, l---arabinitol, arabinitol, l, 2s,4s-pentane-1,2,3,4,5-pentol, l--arabitol
PubChem CID 439255
ChEBI CHEBI:18403
SMILES OCC(O)C(O)C(O)CO

Specifications

Melting Point 99°C to 103°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 100 g
Beilstein 01, 531
Merck Index 10, 784
Specific Rotation Condition −12° (20°C c=2 borax soln 2%)
Specific Rotation -12
IUPAC Name (2S,4S)-pentane-1,2,3,4,5-pentol
Formula Weight 152.15
Percent Purity 99%
Physical Form Powder
Chemical Name or Material L(-)-Arabitol
Show More Show Less
EINECSNumber 231-582-6

RUO – Research Use Only