missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Arabinose 98.0+%, TCI America™

$50.33 - $262.83

Chemical Identifiers

CAS 5328-37-0
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00135866
InChI Key PYMYPHUHKUWMLA-VAYJURFESA-N
Synonym l-+-arabinose, dl-arabinose, 2r,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-arabinose, arabinose, l, unii-b40roo395z, pectinose, pectin sugar, aldehydo-l-arabino-pentose, arabinose,l
PubChem CID 5460291
ChEBI CHEBI:6182
IUPAC Name (2R,3S,4S)-2,3,4,5-tetrahydroxypentanal
SMILES C(C(C(C(C=O)O)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A051525G
View Documents
TCI America
A051525G
25 g N/A
Only null left
 
A0515250G
View Documents
TCI America
A0515250G
250 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

5328-37-0
150.13
PYMYPHUHKUWMLA-VAYJURFESA-N
5460291
(2R,3S,4S)-2,3,4,5-tetrahydroxypentanal
C5H10O5
MFCD00135866
l-+-arabinose, dl-arabinose, 2r,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-arabinose, arabinose, l, unii-b40roo395z, pectinose, pectin sugar, aldehydo-l-arabino-pentose, arabinose,l
CHEBI:6182
C(C(C(C(C=O)O)O)O)O

Specifications

5328-37-0
White
MFCD00135866
l-+-arabinose, dl-arabinose, 2r,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-arabinose, arabinose, l, unii-b40roo395z, pectinose, pectin sugar, aldehydo-l-arabino-pentose, arabinose,l
C(C(C(C(C=O)O)O)O)O
150.13
CHEBI:6182
≥98.0% (GC)
L-(+)-Arabinose
156°C
C5H10O5
25 g
PYMYPHUHKUWMLA-VAYJURFESA-N
(2R,3S,4S)-2,3,4,5-tetrahydroxypentanal
5460291
150.13
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'