missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-(+)-Arabinose, 99%

Catalog No. AAA1192118
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1192118 50 g
AAA1192130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1192118 Supplier Thermo Scientific Chemicals Supplier No. A1192118

CAS: 87-72-9 | C5H10O5 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
L-(+)-Arabinose is used to identify and differentiate and characterize pentose sugar isomerase. It is also used as the carbon source in microbial culture. It is involved in the bioproduction of L-ribose. It finds application in the production of savory reaction flavors. Its derivatives are used in the development of antiviral agents such as nucleoside. It acts as a base component of hemicellulose and pectin.

Solubility
Soluble in water and glycerol. Insoluble in alcohol.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 87-72-9
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00067709
InChI Key SRBFZHDQGSBBOR-VVZXFQNISA-N
Synonym beta-d-arabinopyranose, 2r,3s,4r,5r-oxane-2,3,4,5-tetrol, pentopyranose #, .beta.-d-arabinopyranose, b-arabinopyranose, beta-arabinopyranose 9ci, methyl, a-d-arabinopyranoside, beta-d-arabinopyranose 9ci, 2r,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol, 3s,2r,4r,5r-2h-3,4,5,6-tetrahydropyran-2,3,4,5-tetraol
PubChem CID 25245970
IUPAC Name (4S,5S)-oxane-2,3,4,5-tetrol
SMILES C1C(C(C(C(O1)O)O)O)O

Specifications

Melting Point 155°C to 160°C
Assay Percent Range 99%
Quantity 50 g
Beilstein 1723085
Solubility Information Soluble in water and glycerol. Insoluble in alcohol.
Formula Weight 150.13
Chemical Name or Material L-(+)-Arabinose
EINECSNumber 201-767-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only