missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Arabinose, 99+%

CAS: 87-72-9 | C5H10O5 | 150.13 g/mol

Supplier:  Thermo Scientific Chemicals 104981000

 View more versions of this product

Catalog No. AC104981000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L(+)-Arabinose
155.0°C to 160.0°C
Authentic
Plastic bottle
100 g
10,783
+ 103.00
(10% in water) Clear colorless
C1C(C(C(C(O1)O)O)O)O
150.13
150.13
Fine Crystalline Powder
87-72-9
White
99% min. (ELSD) (HPLC)
C5H10O5
31,32
+103.00 (20.00°C c=5,H2O,24H)
beta-d-arabinopyranose, 3s,2r,4r,5r-2h-3,4,5,6-tetrahydropyran-2,3,4,5-tetraol, pentopyranose #, pentopyranose #, b-arabinopyranose, b-arabinopyranose, beta-arabinopyranose 9ci, methyl, beta-d-arabinopyranose 9ci, beta-d-arabinopyranose 9ci, 2r,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol
SRBFZHDQGSBBOR-VVZXFQNISA-N
(4S,5S)-oxane-2,3,4,5-tetrol
25245970
99+%

Chemical Identifiers

87-72-9
150.13
beta-d-arabinopyranose, 3s,2r,4r,5r-2h-3,4,5,6-tetrahydropyran-2,3,4,5-tetraol, pentopyranose #, pentopyranose #, b-arabinopyranose, b-arabinopyranose, beta-arabinopyranose 9ci, methyl, beta-d-arabinopyranose 9ci, beta-d-arabinopyranose 9ci, 2r,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol
(4S,5S)-oxane-2,3,4,5-tetrol
C5H10O5
SRBFZHDQGSBBOR-VVZXFQNISA-N
25245970
C1C(C(C(C(O1)O)O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 201-767-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only