missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Arabinose, 99+%

Catalog No. AC104985000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104985000 500 g Plastic bottle
AC104980250 25 g Plastic bottle
AC104981000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC104985000 Supplier Thermo Scientific Chemicals Supplier No. 104985000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 87-72-9 | C5H10O5 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87-72-9
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
InChI Key SRBFZHDQGSBBOR-VVZXFQNISA-N
Synonym beta-d-arabinopyranose, 2r,3s,4r,5r-oxane-2,3,4,5-tetrol, pentopyranose #, .beta.-d-arabinopyranose, b-arabinopyranose, beta-arabinopyranose 9ci, methyl, a-d-arabinopyranoside, beta-d-arabinopyranose 9ci, 2r,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol, 3s,2r,4r,5r-2h-3,4,5,6-tetrahydropyran-2,3,4,5-tetraol
PubChem CID 25245970
IUPAC Name (4S,5S)-oxane-2,3,4,5-tetrol
SMILES C1C(C(C(C(O1)O)O)O)O

Specifications

Melting Point 155.0°C to 160.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 31,32
Merck Index 10,783
Specific Rotation Condition +103.00 (20.00°C c=5,H2O,24H)
Specific Rotation + 103.00
Solubility Information (10% in water) Clear colorless
Formula Weight 150.13
Percent Purity 99+%
Physical Form Fine Crystalline Powder
Chemical Name or Material L(+)-Arabinose
Show More Show Less
EINECSNumber 201-767-6

RUO – Research Use Only