missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Arabinose, 99%, for biochemistry

CAS: 87-72-9 | C5H10O5 | 150.13 g/mol

$168.70 - $449.37

Chemical Identifiers

CAS 87-72-9
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
InChI Key SRBFZHDQGSBBOR-VVZXFQNISA-N
Synonym beta-d-arabinopyranose, 3s,2r,4r,5r-2h-3,4,5,6-tetrahydropyran-2,3,4,5-tetraol, pentopyranose #, pentopyranose #, .beta.-d-arabinopyranose, b-arabinopyranose, methyl, a-d-arabinopyranoside, beta-d-arabinopyranose 9ci, 2r,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol, 2r,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol
PubChem CID 25245970
IUPAC Name (4S,5S)-oxane-2,3,4,5-tetrol
SMILES C1C(C(C(C(O1)O)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC365180250
SDS View Documents
Thermo Scientific Chemicals
365180250
25 g
Each for $168.70
Only null left
 
AC365181000
SDS View Documents
Thermo Scientific Chemicals
365181000
100 g
Each for $449.37
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

87-72-9
150.13
beta-d-arabinopyranose, 3s,2r,4r,5r-2h-3,4,5,6-tetrahydropyran-2,3,4,5-tetraol, pentopyranose #, pentopyranose #, .beta.-d-arabinopyranose, b-arabinopyranose, methyl, a-d-arabinopyranoside, beta-d-arabinopyranose 9ci, 2r,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol, 2r,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol
(4S,5S)-oxane-2,3,4,5-tetrol
C5H10O5
SRBFZHDQGSBBOR-VVZXFQNISA-N
25245970
C1C(C(C(C(O1)O)O)O)O

Specifications

87-72-9
0.3% max
C5H10O5
31,32
+ 103.00
(10% in water) Clear colorless
C1C(C(C(C(O1)O)O)O)O
150.13
150.13
Biochemistry
155.0°C to 160.0°C
98.5% min. (ELSD) (HPLC)
25 g
10,783
beta-d-arabinopyranose, 3s,2r,4r,5r-2h-3,4,5,6-tetrahydropyran-2,3,4,5-tetraol, pentopyranose #, pentopyranose #, b-arabinopyranose, beta-arabinopyranose 9ci, beta-arabinopyranose 9ci, a-d-arabinopyranoside, a-d-arabinopyranoside, 2r,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol, 2r,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol
SRBFZHDQGSBBOR-VVZXFQNISA-N
(4S,5S)-oxane-2,3,4,5-tetrol
25245970
99%
L(+)-Arabinose
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 201-767-6

SDS
Documents

Documents

RUO – Research Use Only