missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Arabinose, 99%, for biochemistry

Catalog No. AC365181000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC365181000 100 g
AC365180250 25 g
AC365185000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC365181000 Supplier Thermo Scientific Chemicals Supplier No. 365181000
Only null left

CAS: 87-72-9 | C5H10O5 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87-72-9
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
InChI Key SRBFZHDQGSBBOR-VVZXFQNISA-N
Synonym beta-d-arabinopyranose, 2r,3s,4r,5r-oxane-2,3,4,5-tetrol, pentopyranose #, .beta.-d-arabinopyranose, b-arabinopyranose, beta-arabinopyranose 9ci, methyl, a-d-arabinopyranoside, beta-d-arabinopyranose 9ci, 2r,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol, 3s,2r,4r,5r-2h-3,4,5,6-tetrahydropyran-2,3,4,5-tetraol
PubChem CID 25245970
IUPAC Name (4S,5S)-oxane-2,3,4,5-tetrol
SMILES C1C(C(C(C(O1)O)O)O)O

Specifications

Melting Point 155.0°C to 160.0°C
Ignition Residue 0.3% max
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Quantity 100 g
Beilstein 31,32
Merck Index 10,783
Specific Rotation + 103.00
Solubility Information (10% in water) Clear colorless
Formula Weight 150.13
Percent Purity 99%
Grade Biochemistry
Chemical Name or Material L(+)-Arabinose
Show More Show Less
EINECSNumber 201-767-6

RUO – Research Use Only