missing translation for 'onlineSavingsMsg'
Learn More

L-alpha-Hydroxyisocaproic acid, 98%

CAS: 13748-90-8 | C6H12O3 | 132.16 g/mol

$102.81 - $102.81

Chemical Identifiers

CAS 13748-90-8
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
InChI Key LVRFTAZAXQPQHI-YFKPBYRVSA-N
Synonym l-leucic acid, s-2-hydroxy-4-methylpentanoic acid, 2s-2-hydroxy-4-methylpentanoic acid, l-2-hydroxy-4-methylvaleric acid, s-leucic acid, s-2-hydroxy-4-methylvaleric acid, l-2-hydroxyisocaproic acid, l-alpha-hydroxyisocaproic acid, s---2-hydroxyisocaproic acid, pentanoic acid, 2-hydroxy-4-methyl-, 2s
PubChem CID 83697
ChEBI CHEBI:44510
IUPAC Name (2S)-2-hydroxy-4-methylpentanoic acid
SMILES CC(C)CC(C(=O)O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC204810010
SDS View Documents
Thermo Scientific Chemicals
204810010
1 g
Each for $102.81
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13748-90-8
132.16
l-leucic acid, s-2-hydroxy-4-methylpentanoic acid, 2s-2-hydroxy-4-methylpentanoic acid, l-2-hydroxy-4-methylvaleric acid, s-leucic acid, s-2-hydroxy-4-methylvaleric acid, l-2-hydroxyisocaproic acid, l-alpha-hydroxyisocaproic acid, s---2-hydroxyisocaproic acid, pentanoic acid, 2-hydroxy-4-methyl-, 2s
CHEBI:44510
CC(C)CC(C(=O)O)O
C6H12O3
LVRFTAZAXQPQHI-YFKPBYRVSA-N
83697
(2S)-2-hydroxy-4-methylpentanoic acid

Specifications

13748-90-8
98%
C6H12O3
1 g
l-leucic acid, s-2-hydroxy-4-methylpentanoic acid, 2s-2-hydroxy-4-methylpentanoic acid, l-2-hydroxy-4-methylvaleric acid, s-leucic acid, s-2-hydroxy-4-methylvaleric acid, l-2-hydroxyisocaproic acid, l-alpha-hydroxyisocaproic acid, s---2-hydroxyisocaproic acid, pentanoic acid, 2-hydroxy-4-methyl-, 2s
CC(C)CC(C(=O)O)O
132.16
CHEBI:44510
98%
78°C to 80°C
Glass Bottle
(CH3)2CHCH2CH(OH)COOH
03,336
LVRFTAZAXQPQHI-YFKPBYRVSA-N
(2S)-2-hydroxy-4-methylpentanoic acid
83697
132.16
L-α-Hydroxyisocaproic acid
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 237-329-6

SDS
Documents

Documents

RUO – Research Use Only