missing translation for 'onlineSavingsMsg'
Learn More

L-Alanyl-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog No. p-7056327
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6518203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6518203 Supplier Thermo Scientific Chemicals Supplier No. H6518203
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3061-90-3
Molecular Formula C12H16N2O3
Molecular Weight (g/mol) 236.271
MDL Number MFCD00066031
InChI Key OMNVYXHOSHNURL-WPRPVWTQSA-N
Synonym ala-phe, h-ala-phe-oh, l-alanyl-l-phenylalanine, alanylphenylalanine, s-2-s-2-aminopropanamido-3-phenylpropanoic acid, l-ala-l-phe, chembl57338, ala-phe-oh, phenylalanine, alanyl, 2-2-amino-propionylamino-3-phenyl-propionic acid
PubChem CID 96814
ChEBI CHEBI:73807
IUPAC Name (2S)-2-[[(2S)-2-aminopropanoyl]amino]-3-phenylpropanoic acid
SMILES CC(C(=O)NC(CC1=CC=CC=C1)C(=O)O)N

Specifications

Beilstein 2381598
Quantity 1 g
Formula Weight 236.27
Percent Purity 95%
Physical Form Solid
Chemical Name or Material L-Alanyl-L-phenylalanine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only