accessibility menu, dialog, popup

UserName

H3CH2NOHNOHO

CAS RN 3061-90-3

IUPAC Name: 2-(2-aminopropanamido)-3-phenylpropanoic acid
Synonyms: 2-(2-aminopropanamido)-3-phenylpropanoic acid
Molecular Weight (g/mol): 236.27
Molecular Formula: C12H16N2O3
InChi Key: OMNVYXHOSHNURL-UHFFFAOYNA-N
SMILES: CC(N)C(=O)NC(CC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)