missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine Methyl Ester Hydrochloride 98.0+%, TCI America™

$44.28 - $138.61

Chemical Identifiers

CAS 2491-20-5
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00063663
InChI Key IYUKFAFDFHZKPI-DOIAFFBONA-N
Synonym l-alanine methyl ester hydrochloride, h-ala-ome.hcl, s-methyl 2-aminopropanoate hydrochloride, methyl l-alaninate hydrochloride, h-ala-ome hcl, alanine methyl ester hydrochloride, methyl 2s-2-aminopropanoate hydrochloride, l-ala-ome hcl, d-alanyl ester hydrochloride, l-alanine methyl ester hcl
PubChem CID 2733257
IUPAC Name methyl (2S)-2-aminopropanoate hydrochloride
SMILES Cl.COC(=O)[C@H](C)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A14665G
View Documents
TCI America
A14665G
5 g
Each for $44.28
Only null left
 
A146625G
View Documents
TCI America
A146625G
25 g
Each for $138.61
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2491-20-5
139.58
IYUKFAFDFHZKPI-DOIAFFBONA-N
2733257
Cl.COC(=O)[C@H](C)N
C4H10ClNO2
MFCD00063663
l-alanine methyl ester hydrochloride, h-ala-ome.hcl, s-methyl 2-aminopropanoate hydrochloride, methyl l-alaninate hydrochloride, h-ala-ome hcl, alanine methyl ester hydrochloride, methyl 2s-2-aminopropanoate hydrochloride, l-ala-ome hcl, d-alanyl ester hydrochloride, l-alanine methyl ester hcl
methyl (2S)-2-aminopropanoate hydrochloride

Specifications

2491-20-5
White-Yellow
MFCD00063663
l-alanine methyl ester hydrochloride, h-ala-ome.hcl, s-methyl 2-aminopropanoate hydrochloride, methyl l-alaninate hydrochloride, h-ala-ome hcl, alanine methyl ester hydrochloride, methyl 2s-2-aminopropanoate hydrochloride, l-ala-ome hcl, d-alanyl ester hydrochloride, l-alanine methyl ester hcl
Cl.COC(=O)[C@H](C)N
139.58
139.58
Crystalline Powder
108°C
C4H10ClNO2
5 g
IYUKFAFDFHZKPI-DOIAFFBONA-N
methyl (2S)-2-aminopropanoate hydrochloride
2733257
≥98.0% (T)
L-Alanine Methyl Ester Hydrochloride
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'