missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine Isopropyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. p-7117978
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
A25885G 5 g
A258825G 25 g
2 options

Catalog No. A25885G

Supplier: TCI America A25885G

Only null left

Chemical Identifiers

CAS 62062-65-1
Molecular Formula C6H14ClNO2
Molecular Weight (g/mol) 167.63
MDL Number MFCD08059709
InChI Key YAQKNCSWDMGPOY-UHFFFAOYNA-N
Synonym (S)-2-Aminopropionic Acid Isopropyl Ester Hydrochloride, Isopropyl (S)-2-Aminopropionate Hydrochloride
PubChem CID 13089459
IUPAC Name propan-2-yl 2-aminopropanoate hydrochloride
SMILES Cl.CC(C)OC(=O)C(C)N

Specifications

Melting Point 85°C
Color White-Yellow
Quantity 5 g
Formula Weight 167.63
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Alanine Isopropyl Ester Hydrochloride