accessibility menu, dialog, popup

UserName

CAS RN 62062-65-1

H3CCH3OOCH3NH2HCl

CAS RN 62062-65-1

IUPAC Name: propan-2-yl 2-aminopropanoate hydrochloride
Synonyms: isopropyl 2-aminopropanoate hydrochloride
Molecular Weight (g/mol): 167.63
Molecular Formula: C6H14ClNO2
InChi Key: YAQKNCSWDMGPOY-UHFFFAOYNA-N
SMILES: Cl.CC(C)OC(=O)C(C)N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results