missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Alanine, Cell Culture Reagent

CAS: 56-41-7 | C3H7NO2 | 89.09 g/mol

Supplier:  Thermo Scientific Chemicals J6027922

 View more versions of this product

Catalog No. AAJ6027922


Only null left

Description

Description

It is a key player in the Glucose-Alanine cycle, which enables the removal of pyruvate and glutamate from muscle to the liver. The Glucose-Alanine cycle aides to conserve ATP in muscle for muscle contraction, while the energy burden of gluconeogenesis is imposed upon the liver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a key player in the Glucose-Alanine cycle, which enables the removal of pyruvate and glutamate from muscle to the liver. The Glucose-Alanine cycle aides to conserve ATP in muscle for muscle contraction, while the energy burden of gluconeogenesis is imposed upon the liver.

Solubility
Soluble in water 166.5 g/L..

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
Specifications

Specifications

∽295°C (decomposition)
56-41-7
C3H7NO2
1720248
l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
QNAYBMKLOCPYGJ-REOHCLBHSA-N
(2S)-2-aminopropanoic acid
100 g
CHEBI:16977
89.09
Powder
L-Alanine
White
MFCD00064410
14,204
Soluble in water 166.5g/L..
C[C@H](N)C(O)=O
89.09
5950
+14° (c=5 in 5N HCl)
Cell Culture Reagent

Chemical Identifiers

56-41-7
89.09
QNAYBMKLOCPYGJ-REOHCLBHSA-N
5950
(2S)-2-aminopropanoic acid
C3H7NO2
MFCD00064410
l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
CHEBI:16977
C[C@H](N)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-273-8

RTECSNumber : AY2990000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only