missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine, 99%

Catalog No. p-3734442
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102830250 25 g Plastic Bottle
AC102831000 100 g Plastic Bottle
AC102835000 500 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC102830250 Supplier Thermo Scientific Chemicals Supplier No. 102830250
Only null left

CAS: 56-41-7 | C3H7NO2 | 89.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56-41-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064410
InChI Key QNAYBMKLOCPYGJ-REOHCLBHSA-N
Synonym l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
PubChem CID 5950
ChEBI CHEBI:16977
IUPAC Name (2S)-2-aminopropanoic acid
SMILES C[C@H](N)C(O)=O

Specifications

Melting Point 314.0°C to 316.0°C
Ammonium 200ppm max.
pH 5.5 to 7 (2.5% soln. at 25°C)
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Loss on Drying 0.2% max. (105°C, 3 hrs)
Assay Percent Range 99%
Other Amino Acids 0.5% max. (TLC)
Packaging Plastic Bottle
Linear Formula CH3CH(NH2)CO2H
Specific Rotation + 14.50
Beilstein 04, 381
Sulfate 300ppm max.
Transmittance 95% min. (c=10, H2O, 430nm) 1cm cell
Merck Index 15, 197
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 166.5g/l in water (25°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and organic solvents
Quantity 25 g
Formula Weight 89.09
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Alanine
Show More Show Less

RUO – Research Use Only