missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Alanine, 99%, specified according to the requirements of Ph.Eur.
CAS: 56-41-7 | C3H7NO2 | 89.09 g/mol
Supplier: Thermo Scientific Chemicals 451850025
| Quantity | 2.5 kg |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C3H7NO2 | |
| MFCD00064410 | |
| l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid | |
| CHEBI:16977 | |
| C[C@H](N)C(O)=O |
Specifications
| 314.0°C to 316.0°C | |
| specified according to the requirements of Ph.Eur. | |
| 99% | |
| CH3CH(NH2)CO2H | |
| + 14.50 | |
| Solubility in water: 166.5g/l in water (25°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and organic solvents | |
| C[C@H](N)C(O)=O | |
| 89.09 | |
| 5950 | |
| 89.09 |
| L-Alanine | |
| 56-41-7 | |
| C3H7NO2 | |
| MFCD00064410 | |
| l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid | |
| QNAYBMKLOCPYGJ-REOHCLBHSA-N | |
| (2S)-2-aminopropanoic acid | |
| 2.5 kg | |
| CHEBI:16977 | |
| 99% |