missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Alanine, 99%, MP Biomedicals™
A nonessential amino acid that plays a key role in the glucose-alanine cycle between muscle tissue and the liver. In amino acid-degrading tissues such as muscle, amino groups are pooled as glutamate by transamination reactions
Supplier: MP Biomedicals Inc 0210028705
| Quantity | 5 kg |
|---|
Chemical Identifiers
| C3H7NO2 | |
| MFCD00064410 | |
| l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid | |
| CHEBI:16977 | |
| C[C@H](N)C(O)=O |
Specifications
| L-Alanine | |
| White | |
| C3H7NO2 | |
| l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid | |
| C[C@H](N)C(O)=O | |
| (2S)-2-aminopropanoic acid | |
| 5 kg | |
| CHEBI:16977 | |
| Powder |
| 56-41-7 | |
| 0.99 | |
| MFCD00064410 | |
| QNAYBMKLOCPYGJ-REOHCLBHSA-N | |
| 1.40g/mL | |
| 89.09 | |
| 5950 | |
| 89.1 |
Safety and Handling
EINECSNumber : 200-273-8