missing translation for 'onlineSavingsMsg'
Learn More

L-5-Hydroxytryptophan hydrate, 98%, Thermo Scientific Chemicals

$33.59 - $70.51

Chemical Identifiers

CAS 9-8-4350
MDL Number MFCD00064341
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA1395401
View Documents Promotion Details
N/A 0.25 g
Each for $33.59
Only null left
 
AAA1395403
View Documents Promotion Details
N/A 1 g
Each for $70.51
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9-8-4350
MFCD00064341

Specifications

258°C (decomposition)
C10H16O4
UN2811
14,4847
YQPCHPBGAALCRT-UHFFFAOYSA-N
2-[1-(carboxymethyl)cyclohexyl]acetic acid
78061
−33° (c=1 in Water)
L-5-Hydroxytryptophan hydrate
9-8-4350
MFCD00064341
88200
1,1-cyclohexanediacetic acid, 1,1-cyclohexanediaceticacid, cyclohexanediacetic acid, 2-1-carboxymethyl cyclohexyl acetic acid, 1,1-cyclohexane diacetic acid, 1-carboxymethyl cyclohexyl acetic acid, 2,2'-cyclohexane-1,1-diyl diacetic acid, 2-carboxymethyl cyclohexyl acetic acid, gabapentin impurity 2, pubchem13645
C1CCC(CC1)(CC(=O)O)CC(=O)O
200.23
220.23 (Anhydrous)
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 224-411-1

RTECSNumber : YN7110000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only