missing translation for 'onlineSavingsMsg'
Learn More

L-5-Hydroxytryptophan hydrate, 98%, Thermo Scientific Chemicals

Catalog No. AAA1395401
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1395401 0.25 g
AAA1395403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1395401 Supplier Thermo Scientific Chemicals Supplier No. A1395401

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9-8-4350
MDL Number MFCD00064341

Specifications

Melting Point 258°C (decomposition)
Molecular Formula C10H16O4
UN Number UN2811
Beilstein 88200
Merck Index 14,4847
Synonym 1,1-cyclohexanediacetic acid, 1,1-cyclohexanediaceticacid, cyclohexanediacetic acid, 2-1-carboxymethyl cyclohexyl acetic acid, 1,1-cyclohexane diacetic acid, 1-carboxymethyl cyclohexyl acetic acid, 2,2'-cyclohexane-1,1-diyl diacetic acid, 2-carboxymethyl cyclohexyl acetic acid, gabapentin impurity 2, pubchem13645
InChI Key YQPCHPBGAALCRT-UHFFFAOYSA-N
SMILES C1CCC(CC1)(CC(=O)O)CC(=O)O
IUPAC Name 2-[1-(carboxymethyl)cyclohexyl]acetic acid
Quantity 0.25 g
Molecular Weight (g/mol) 200.23
PubChem CID 78061
Formula Weight 220.23 (Anhydrous)
Optical Rotation −33° (c=1 in Water)
Percent Purity 98%
Chemical Name or Material L-5-Hydroxytryptophan hydrate
Show More Show Less
Hazard Category H301
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 224-411-1
RTECSNumber YN7110000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only