missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-(+)-2-Phenylglycine, 98+%
CAS: 2935-35-5 | C8H9NO2 | 151.165 g/mol
$69.21 - $554.30
Chemical Identifiers
| CAS | 2935-35-5 |
|---|---|
| Molecular Formula | C8H9NO2 |
| Molecular Weight (g/mol) | 151.165 |
| MDL Number | MFCD00064403 |
| InChI Key | ZGUNAGUHMKGQNY-ZETCQYMHSA-N |
| Synonym | h-phg-oh, l-phenylglycine, s-2-amino-2-phenylacetic acid, l-+-alpha-phenylglycine, l-2-phenylglycine, l-+-2-phenylglycine, s-phenylglycine, phenylglycine, 2s-2-amino-2-phenylacetic acid, 2s-amino phenyl acetic acid |
| PubChem CID | 99291 |
| ChEBI | CHEBI:439819 |
| IUPAC Name | (2S)-2-amino-2-phenylacetic acid |
| SMILES | C1=CC=C(C=C1)C(C(=O)O)N |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
AAA1936014
|
Thermo Scientific Chemicals
A1936014 |
25 g |
Each for $69.21
|
|
|||||
|
AAA1936022
|
Thermo Scientific Chemicals
A1936022 |
100 g |
Each for $154.82
|
|
|||||
|
AAA1936036
|
Thermo Scientific Chemicals
A1936036 |
500 g |
Each for $554.30
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2935-35-5 | |
| 151.165 | |
| ZGUNAGUHMKGQNY-ZETCQYMHSA-N | |
| 99291 | |
| (2S)-2-amino-2-phenylacetic acid |
| C8H9NO2 | |
| MFCD00064403 | |
| h-phg-oh, l-phenylglycine, s-2-amino-2-phenylacetic acid, l-+-alpha-phenylglycine, l-2-phenylglycine, l-+-2-phenylglycine, s-phenylglycine, phenylglycine, 2s-2-amino-2-phenylacetic acid, 2s-amino phenyl acetic acid | |
| CHEBI:439819 | |
| C1=CC=C(C=C1)C(C(=O)O)N |
Specifications
| 288°C (decomposition) | |
| C8H9NO2 | |
| 2208675 | |
| h-phg-oh, l-phenylglycine, s-2-amino-2-phenylacetic acid, l-+-alpha-phenylglycine, l-2-phenylglycine, l-+-2-phenylglycine, s-phenylglycine, phenylglycine, 2s-2-amino-2-phenylacetic acid, 2s-amino phenyl acetic acid | |
| C1=CC=C(C=C1)C(C(=O)O)N | |
| 25 g | |
| 99291 | |
| +156° (c=1 in 1N HCl) | |
| ≥98% |
| 2935-35-5 | |
| MFCD00064403 | |
| 14,7291 | |
| ZGUNAGUHMKGQNY-ZETCQYMHSA-N | |
| (2S)-2-amino-2-phenylacetic acid | |
| 151.165 | |
| CHEBI:439819 | |
| 151.17 | |
| L-(+)-2-Phenylglycine |
Safety and Handling
EINECSNumber : 220-909-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only