missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-(+)-2-Phenylglycine, 98+%

Catalog No. aaa1936014
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1936014 25 g
AAA1936022 100 g
AAA1936036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1936014 Supplier Thermo Scientific Chemicals Supplier No. A1936014
Only null left

CAS: 2935-35-5 | C8H9NO2 | 151.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2935-35-5
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00064403
InChI Key ZGUNAGUHMKGQNY-ZETCQYMHSA-N
Synonym h-phg-oh, l-phenylglycine, s-2-amino-2-phenylacetic acid, l-+-alpha-phenylglycine, l-2-phenylglycine, l-+-2-phenylglycine, s-phenylglycine, phenylglycine, 2s-2-amino-2-phenylacetic acid, 2s-amino phenyl acetic acid
PubChem CID 99291
ChEBI CHEBI:439819
IUPAC Name (2S)-2-amino-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)N

Specifications

Melting Point 288°C (decomposition)
Beilstein 2208675
Merck Index 14,7291
Quantity 25 g
Formula Weight 151.17
Optical Rotation +156° (c=1 in 1N HCl)
Percent Purity ≥98%
Chemical Name or Material L-(+)-2-Phenylglycine
EINECSNumber 220-909-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only