missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-2-Aminobutyric acid, 98+%

Catalog No. AAL1398414
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1398414 25 g
AAL1398403 1 g
AAL1398406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1398414 Supplier Thermo Scientific Chemicals Supplier No. L1398414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1492-24-6 | C4H9NO2 | 103.121 g/mol

L-(+)-2-Aminobutyric acid is used in the biosynthesis of nonribosomal peptides. It acts as a receptor antagonist. It is also used as a chiral reagent. Further, it is used in the determination of substrate of glutamyl cysteine acid synthase. In addition to this, it is also utilized as a drug intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(+)-2-Aminobutyric acid is used in the biosynthesis of nonribosomal peptides. It acts as a receptor antagonist. It is also used as a chiral reagent. Further, it is used in the determination of substrate of glutamyl cysteine acid synthase. In addition to this, it is also utilized as a drug intermediate.

Solubility
Soluble in water and acetic acid. Slightly soluble in alcohol and ether.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1492-24-6
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00064415
InChI Key QWCKQJZIFLGMSD-VKHMYHEASA-N
Synonym l-2-aminobutyric acid, s-2-aminobutanoic acid, 2s-2-aminobutanoic acid, h-abu-oh, l-alpha-aminobutyric acid, s-+-2-aminobutyric acid, l-+-2-aminobutyric acid, --2-aminobutyric acid, s-2-aminobutyric acid, s-2-amino-butyric acid
PubChem CID 80283
ChEBI CHEBI:35619
IUPAC Name (2S)-2-aminobutanoic acid
SMILES CCC(C(=O)O)N

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 1720935
Merck Index 14,428
Solubility Information Soluble in water and acetic acid. Slightly soluble in alcohol and ether.
Optical Rotation +21° (c=4.8 in 6N HCl)
Formula Weight 103.12
Percent Purity ≥98%
Chemical Name or Material L-(+)-2-Aminobutyric acid
Hazard Category Warning
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 216-083-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only